Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLSCR1
Entrez Gene ID
100125943
Gene Name
phospholipid scramblase 1
Gene Aliases
-
Chromosome Location
Chr.1: 122001997 - 122092791 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105506.2 NP_001098976.2 3 - 4 283 146
GenBank mRNA BC149269.1 - 3 - 4 284 146

View Full Product Details