Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IZUMO3
Entrez Gene ID
786878
Gene Name
IZUMO family member 3
Gene Aliases
-
Chromosome Location
Chr.8: 19634212 - 19637417 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105642.1 NP_001099112.1 1 - 2 464 65
GenBank mRNA BC147884.1 - 1 - 2 464 65

View Full Product Details