Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM77
Entrez Gene ID
100126826
Gene Name
tripartite motif containing 77
Gene Aliases
RNF18, TRIM49, TRIM77P
Chromosome Location
Chr.29: 5190831 - 5211296 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111262.2 NP_001104732.1 3 - 4 647 130
GenBank mRNA EF446903.1 - 3 - 4 648 130

View Full Product Details