Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HBB
Entrez Gene ID
280813
Gene Name
hemoglobin, beta
Gene Aliases
GLNA1
Chromosome Location
Chr.15: 48362354 - 48363996 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173917.2 NP_776342.1 2 - 3 368 151
GenBank mRNA DY466682.1 - 2 - 3 326 151

View Full Product Details