Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAG4
Entrez Gene ID
337898
Gene Name
pregnancy-associated glycoprotein 4
Gene Aliases
-
Chromosome Location
Chr.29: 38217572 - 38226738 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176615.2 NP_788788.1 3 - 4 358 120
GenBank mRNA AF020506.1 - 3 - 4 358 120

View Full Product Details