Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DOLK
Entrez Gene ID
526844
Gene Name
dolichol kinase
Gene Aliases
TMEM15
Chromosome Location
Chr.11: 99443608 - 99445710 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035365.3 NP_001030442.2 1 - 1 143 150
GenBank mRNA BC126569.1 - 1 - 1 144 150

View Full Product Details