Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRS1
Entrez Gene ID
510956
Gene Name
ribosome biogenesis regulator 1 homolog
Gene Aliases
-
Chromosome Location
Chr.14: 30687060 - 30688825 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075481.2 NP_001068949.1 1 - 1 1548 119
GenBank mRNA BC112636.1 - 1 - 1 1550 119

View Full Product Details