Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IER2
Entrez Gene ID
525380
Gene Name
immediate early response 2
Gene Aliases
-
Chromosome Location
Chr.7: 12370002 - 12438331 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075871.1 NP_001069339.1 1 - 1 552 81
GenBank mRNA BC102465.1 - 1 - 1 552 81

View Full Product Details