Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
SLC66A2
Entrez Gene ID
533304
Gene Name
solute carrier family 66 member 2
Gene Aliases
PQLC1
Chromosome Location
Chr.24: 569420 - 610884 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076893.1 NP_001070361.1 5 - 5 2250 138
GenBank mRNA BC120242.1 - 5 - 5 2250 138

View Full Product Details