The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001078105.1 | NP_001071573.1 | 3 - 3 | 1952 | 146 |
GenBank mRNA | BC114104.1 | - | 3 - 3 | 1952 | 146 |