Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
SSLP1
Entrez Gene ID
100125878
Gene Name
secreted seminal-vesicle Ly-6 protein 1
Gene Aliases
SOLD1, SSLP-1
Chromosome Location
Chr.29: 29084974 - 29092471 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105478.1 NP_001098948.1 1 - 1 45 141
GenBank mRNA BC149580.1 - 1 - 1 45 141
EU846101.1 - 1 - 1 45 141

View Full Product Details