Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NXPH2
Entrez Gene ID
281956
Gene Name
neurexophilin 2
Gene Aliases
-
Chromosome Location
Chr.2: 58351070 - 58477701 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174406.2 NP_776831.1 2 - 2 1154 106
GenBank mRNA L27868.1 - 2 - 2 1154 106

View Full Product Details