Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TAP
Entrez Gene ID
286837
Gene Name
tracheal antimicrobial peptide
Gene Aliases
-
Chromosome Location
Chr.27: 6013830 - 6015648 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174776.1 NP_777201.1 2 - 2 151 139
GenBank mRNA AF014106.1 - 2 - 2 151 139

View Full Product Details