Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GRP
Entrez Gene ID
615323
Gene Name
gastrin releasing peptide
Gene Aliases
-
Chromosome Location
Chr.24: 58203569 - 58216205 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101239.1 NP_001094709.1 1 - 1 153 135
GenBank mRNA BC149933.1 - 1 - 1 153 135

View Full Product Details