Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRPV4
Entrez Gene ID
540259
Gene Name
transient receptor potential cation channel subfamily V member 4
Gene Aliases
-
Chromosome Location
Chr.17: 63410069 - 63448313 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192385.3 NP_001179314.1 6 - 7 1333 59

View Full Product Details