Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHCHD7
Entrez Gene ID
616237
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 7
Gene Aliases
-
Chromosome Location
Chr.14: 23376149 - 23383137 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130759.2 NP_001124231.1 3 - 4 224 68
GenBank mRNA BC118483.1 - 3 - 4 225 68

View Full Product Details