Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MATN1
Entrez Gene ID
512059
Gene Name
matrilin 1
Gene Aliases
-
Chromosome Location
Chr.2: 122848310 - 122860863 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143866.1 NP_001137338.1 6 - 7 1409 77

View Full Product Details