Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SHISA9
Entrez Gene ID
100139490
Gene Name
shisa family member 9
Gene Aliases
CKAMP44
Chromosome Location
Chr.25: 11633245 - 12158399 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145203.1 NP_001138675.2 4 - 5 1073 72
GenBank mRNA EH181025.1 - 4 - 5 409 72

View Full Product Details