Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOS1AP
Entrez Gene ID
539686
Gene Name
nitric oxide synthase 1 adaptor protein
Gene Aliases
-
Chromosome Location
Chr.3: 7126370 - 7467854 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146219.1 NP_001139691.1 6 - 7 731 78

View Full Product Details