Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SHBG
Entrez Gene ID
404182
Gene Name
sex hormone binding globulin
Gene Aliases
-
Chromosome Location
Chr.19: 27345874 - 27350673 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098858.2 NP_001092328.1 2 - 3 331 63
GenBank mRNA AY443491.1 - 1 - 2 91 63
BC142435.1 - 2 - 3 332 63

View Full Product Details