Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CKAP2L
Entrez Gene ID
507498
Gene Name
cytoskeleton associated protein 2 like
Gene Aliases
-
Chromosome Location
Chr.11: 46451749 - 46474168 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098901.2 NP_001092371.1 2 - 3 131 76
GenBank mRNA BC142323.1 - 2 - 3 132 76

View Full Product Details