Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NGFR
Entrez Gene ID
353110
Gene Name
nerve growth factor receptor
Gene Aliases
-
Chromosome Location
Chr.19: 37093168 - 37112123 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102478.2 NP_001095948.1 5 - 6 1114 67
GenBank mRNA BC142255.1 - 5 - 6 1115 67

View Full Product Details