Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TWF2
Entrez Gene ID
282024
Gene Name
twinfilin actin binding protein 2
Gene Aliases
PTK9L
Chromosome Location
Chr.22: 48665271 - 48674428 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105324.2 NP_001098794.1 4 - 5 513 70
GenBank mRNA BC133441.1 - 4 - 5 515 70
BC149594.1 - 4 - 5 528 70

View Full Product Details