Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SKINT1
Entrez Gene ID
528184
Gene Name
selection and upkeep of intraepithelial T cells 1
Gene Aliases
-
Chromosome Location
Chr.3: 98066623 - 98090010 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127316.1 NP_001120788.1 1 - 2 77 65
GenBank mRNA EU650233.1 - 1 - 2 77 65

View Full Product Details