The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127440.1 | NP_001120912.1 | 2 - 3 | 1155 | 77 |
GenBank mRNA | AJ936934.2 | - | 2 - 3 | 1155 | 77 |