Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TGIF1
Entrez Gene ID
510050
Gene Name
TGFB induced factor homeobox 1
Gene Aliases
-
Chromosome Location
Chr.24: 37498883 - 37535210 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128499.1 NP_001121971.1 1 - 2 293 93
GenBank mRNA BC103302.2 - 1 - 2 293 93

View Full Product Details