Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KMT2E
Entrez Gene ID
539121
Gene Name
lysine methyltransferase 2E (inactive)
Gene Aliases
MLL5
Chromosome Location
Chr.4: 46202169 - 46293391 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082451.3 NP_001075920.2 24 - 25 4355 75

View Full Product Details