Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
DDAH1
Entrez Gene ID
537391
Gene Name
dimethylarginine dimethylaminohydrolase 1
Gene Aliases
-
Chromosome Location
Chr.3: 58618306 - 58775122 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102201.2 NP_001095671.1 2 - 3 506 83
GenBank mRNA BC107532.1 - 2 - 3 505 83
BC149998.1 - 2 - 3 507 83

View Full Product Details