Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
APOF
Entrez Gene ID
539595
Gene Name
apolipoprotein F
Gene Aliases
-
Chromosome Location
Chr.5: 56979557 - 56994574 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128502.2 NP_001121974.1 1 - 2 65 78
GenBank mRNA BC118363.1 - 1 - 2 66 78

View Full Product Details