Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPC3
Entrez Gene ID
100297753
Gene Name
two pore channel 3
Gene Aliases
-
Chromosome Location
Chr.11: 1621153 - 1675048 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161651.1 NP_001155123.1 8 - 9 912 89

View Full Product Details