Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYB5B
Entrez Gene ID
506370
Gene Name
cytochrome b5 type B
Gene Aliases
-
Chromosome Location
Chr.18: 36548148 - 36569868 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163782.2 NP_001157254.1 1 - 2 192 67
GenBank mRNA BC120115.1 - 1 - 2 193 67

View Full Product Details