Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CDYL2
Entrez Gene ID
536901
Gene Name
chromodomain Y like 2
Gene Aliases
-
Chromosome Location
Chr.18: 7389325 - 7553534 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163785.1 NP_001157257.1 5 - 6 1245 73
GenBank mRNA EF690283.1 - 5 - 6 1245 73

View Full Product Details