Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VNN2
Entrez Gene ID
534929
Gene Name
vanin 2
Gene Aliases
-
Chromosome Location
Chr.9: 70886776 - 70916016 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163920.1 NP_001157392.1 1 - 2 211 106
GenBank mRNA BT030550.1 - 1 - 2 211 106

View Full Product Details