Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL26
Entrez Gene ID
782075
Gene Name
interleukin 26
Gene Aliases
-
Chromosome Location
Chr.5: 45548403 - 45567433 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205424.1 NP_001192353.1 4 - 5 428 100

View Full Product Details