Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPIB
Entrez Gene ID
784460
Gene Name
Spi-B transcription factor
Gene Aliases
B42b-like
Chromosome Location
Chr.18: 56573095 - 56580060 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205444.3 NP_001192373.3 3 - 4 127 92

View Full Product Details