Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDCD7
Entrez Gene ID
784973
Gene Name
programmed cell death 7
Gene Aliases
-
Chromosome Location
Chr.10: 12071022 - 12082676 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205449.1 NP_001192378.1 3 - 4 1278 76

View Full Product Details