Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRXCR2
Entrez Gene ID
787763
Gene Name
glutaredoxin and cysteine rich domain containing 2
Gene Aliases
-
Chromosome Location
Chr.7: 57065186 - 57086707 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205458.2 NP_001192387.1 1 - 2 333 126

View Full Product Details