Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRG3
Entrez Gene ID
539977
Gene Name
neuregulin 3
Gene Aliases
-
Chromosome Location
Chr.28: 36896385 - 38133747 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205478.1 NP_001192407.1 7 - 8 1427 75

View Full Product Details