Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHOQ
Entrez Gene ID
100139988
Gene Name
ras homolog family member Q
Gene Aliases
-
Chromosome Location
Chr.11: 28991537 - 29031801 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205498.1 NP_001192427.1 1 - 2 419 70

View Full Product Details