Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZKSCAN1
Entrez Gene ID
100140434
Gene Name
zinc finger with KRAB and SCAN domains 1
Gene Aliases
-
Chromosome Location
Chr.25: 36399127 - 36418859 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205510.3 NP_001192439.1 1 - 2 504 62

View Full Product Details