Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM83G
Entrez Gene ID
100139162
Gene Name
family with sequence similarity 83 member G
Gene Aliases
-
Chromosome Location
Chr.19: 34229739 - 34260824 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205516.1 NP_001192445.1 3 - 4 802 78

View Full Product Details