Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MDFI
Entrez Gene ID
286808
Gene Name
MyoD family inhibitor
Gene Aliases
-
Chromosome Location
Chr.23: 15457487 - 15474727 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205537.1 NP_001192466.1 1 - 2 201 110

View Full Product Details