Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PUS7L
Entrez Gene ID
505464
Gene Name
pseudouridine synthase 7 like
Gene Aliases
-
Chromosome Location
Chr.5: 36708358 - 36729838 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205568.3 NP_001192497.2 2 - 3 1067 115

View Full Product Details