Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CLEC4G
Entrez Gene ID
507332
Gene Name
C-type lectin domain family 4 member G
Gene Aliases
-
Chromosome Location
Chr.7: 16561211 - 16564668 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205606.3 NP_001192535.3 5 - 6 426 56

View Full Product Details