Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZBTB49
Entrez Gene ID
507385
Gene Name
zinc finger and BTB domain containing 49
Gene Aliases
ZNF509
Chromosome Location
Chr.6: 104792357 - 104819010 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205607.1 NP_001192536.1 5 - 6 1454 90

View Full Product Details