Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C14H8orf76
Entrez Gene ID
539014
Gene Name
chromosome 14 C8orf76 homolog
Gene Aliases
-
Chromosome Location
Chr.14: 16566024 - 16597527 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205614.2 NP_001192543.1 2 - 3 620 90
GenBank mRNA BC149302.1 - 2 - 3 214 90

View Full Product Details