Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC39A8
Entrez Gene ID
508193
Gene Name
solute carrier family 39 member 8
Gene Aliases
-
Chromosome Location
Chr.6: 22459057 - 22542658 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205630.1 NP_001192559.1 3 - 4 764 99

View Full Product Details