Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAS2L2
Entrez Gene ID
539169
Gene Name
growth arrest specific 2 like 2
Gene Aliases
-
Chromosome Location
Chr.19: 14621309 - 14629037 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205642.1 NP_001192571.1 4 - 5 832 113

View Full Product Details