Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DSCC1
Entrez Gene ID
509563
Gene Name
DNA replication and sister chromatid cohesion 1
Gene Aliases
-
Chromosome Location
Chr.14: 81263986 - 81276602 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205657.2 NP_001192586.2 3 - 4 471 97
GenBank mRNA CV981480.1 - 3 - 4 372 97

View Full Product Details