Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH2D3A
Entrez Gene ID
510173
Gene Name
SH2 domain containing 3A
Gene Aliases
-
Chromosome Location
Chr.7: 17732344 - 17747013 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205669.1 NP_001192598.1 4 - 5 555 74
GenBank mRNA BT025405.1 - 4 - 5 554 74

View Full Product Details